Skip to Content
Merck
All Photos(2)

Key Documents

BAH131870000

Poxygrid® bag holder

to hold, 12 x 24 in (30 x 61 cm) bag

Synonym(s):

bag holder

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
€110.00

€110.00


Please contact Customer Service for Availability


Select a Size

Change View
1 EACH
€110.00

About This Item

UNSPSC Code:
41122800
NACRES:
NB.43

€110.00


Please contact Customer Service for Availability

feature

to hold 12 x 24 in (30 x 61 cm) bag

packaging

pack of 1 ea

manufacturer/tradename

Bel-Art H13187-0000

diam. × H

7.3 in. (185 mm) × 20.5 in. (520 mm)

Looking for similar products? Visit Product Comparison Guide

General description

Sturdy wire holder keeps bags open while a separate hooked ring that rests over the top keeps them securely in place

Features and Benefits

  • Bright biohazard orange colored epoxy-coated steel wire
  • Chemical and corrosion-resistant; unaffected by organic solvents, salts, weak acids and weak alkalis
  • Will withstand a temperature range of -223ºC (-369ºF) to 121ºC (250ºF)
  • Autoclavable

Legal Information

Poxygrid is a registered trademark of SP Scienceware, a division of SP

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service