Skip to Content
Merck
All Photos(1)

Key Documents

933171

Sigma-Aldrich

SEM Sample Holder

Four Pin Stub Holder

Synonym(s):

4-pin SEM mount

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41111761
NACRES:
NB.35
Pricing and availability is not currently available.

Looking for similar products? Visit Product Comparison Guide

General description

EMS techware

Application

Used for clamping in scanning electron microscope (SEM). It accommodates four pin type, up to 12.5 (½") surface specimen stubs, with ⅛" (3.1 mm) diameter pin.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service