Skip to Content
Merck
All Photos(2)

Key Documents

930857

Sigma-Aldrich

Ni(bpy)Cl2

greener alternative

Synonym(s):

[Bipyridine]nickel(II) dichloride

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€86.40
500 MG
€269.00

€86.40


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
€86.40
500 MG
€269.00

About This Item

Empirical Formula (Hill Notation):
C10H8Cl2N2Ni
CAS Number:
Molecular Weight:
285.78
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.21

€86.40


Please contact Customer Service for Availability

Request a Bulk Order

form

powder

Quality Level

greener alternative product characteristics

Catalysis
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

greener alternative category

storage temp.

−20°C

SMILES string

Cl[Ni]1([N]2=CC=CC=C2C3=CC=CC=[N]31)Cl

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Green Chemistry. This product has been enhanced for catalytic efficiency. Click here for more information.

Application

Ni(bpy)Cl2 is a nickel catalyst used for a variety of cross-coupling reactions.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alexis L Gabbey et al.
Organic letters, 24(17), 3173-3178 (2022-04-27)
The transition-metal-catalyzed α-arylation of secondary amides remains a synthetic challenge due to the presence of a free N-H bond. We report a strategy to synthesize secondary α-aryl amides via a Ni-catalyzed reductive arylation of redox-active N-hydroxyphthalimide (NHP) esters of malonic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service