Skip to Content
Merck
All Photos(2)

Documents

911844

Sigma-Aldrich

(Me4Phen)NiCl2

Synonym(s):

3,4,7,8-Tetramethyl-1,10-phenanthroline nickel (II) dichloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H16Cl2N2Ni
CAS Number:
Molecular Weight:
365.91
UNSPSC Code:
12352200

form

powder

reaction suitability

reagent type: catalyst
reaction type: Cross Couplings

mp

231 °C (decomp.)

Application

(Me4Phen)NiCl2 is a Ni catalyst used for a variety of cross-coupling reactions including the cross-electrophile coupling of benzyl alcohols and aryl halides.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Muta. 2 - Repr. 2 - Resp. Sens. 1 - STOT RE 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Takuya Suga et al.
Organic letters, 20(24), 7846-7850 (2018-12-12)
A nickel-catalyzed cross-electrophile coupling reaction between benzyl alcohols and aryl halides has been developed using a homolytic C-O bond cleavage protocol that has recently been established. The treatment of a benzyl alcohol and aryl halide with a nickel catalyst and

Articles

Nickel complexes catalyze various synthetic reactions like oxidative addition, C-H activation, and cross-coupling.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service