Skip to Content
Merck
All Photos(1)

Key Documents

929956

Sigma-Aldrich

Lithocholic acid-3,4,23,24-13C4 solution

≥98 atom % 13C, ≥98% (CP), 100 μg/mL in methanol

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
€198.00

€198.00


Please contact Customer Service for Availability

Request more information

Select a Size

Change View
1 ML
€198.00

About This Item

Empirical Formula (Hill Notation):
13C3C21H40O3
Molecular Weight:
379.55
UNSPSC Code:
12352106
NACRES:
NA.13

€198.00


Please contact Customer Service for Availability

Request more information

isotopic purity

≥98 atom % 13C

Quality Level

Assay

≥98% (CP)

concentration

100 μg/mL in methanol

mass shift

M+3

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F

Flash Point(C)

9.7 °C


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service