Skip to Content
Merck
All Photos(1)

Documents

928542

Sigma-Aldrich

24-well Chip

Fluidic 1105, COC

Synonym(s):

Cyclic olefin copolymer, Microfluidic chip

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
42142600
NACRES:
NA.25

description

Microfludic chip x1

Looking for similar products? Visit Product Comparison Guide

Application

The 24-well Chip, Fluidic 1105, COC is ideal for screening and evaluating different parameters of a microfluidic experiment, such as different surface treatments, coating protocols, or reagents/solvents.

Features and Benefits

  • 24 flat-bottom 50 µl wells (open to the top)
  • Can be used with an automated pipetting system
  • Ideal for screening multiple parameters at once
Chip Properties:
  • Mini Luer Interface
  • Material: Cyclic olefin copolymer (COC)
  • 24 flat-bottom wells
  • Well volume: 50 µl

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service