CLS3679
Corning® UV-Transparent Microplates
half area, flat bottom clear, pkg of 25 per bag, non-sterile, lid: no
Synonym(s):
96 multiwell plate, 96 well microplate, 96 well microtiter plate, 96 well plate, EIA/RIA plate, immunoassay plate
Select a Size
€1,210.00
Select a Size
About This Item
flat bottom clear
round clear wells
€1,210.00
material
clear polystyrene plate
flat bottom clear
round clear wells
sterility
non-sterile
feature
lid: no
skirt
half area
packaging
pkg of 25 per bag
manufacturer/tradename
Corning 3679
size
96 wells
well volume
205 μL
well working volume
25-125 μL
color
clear
suitability
suitable for (cell cuture)
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Features and Benefits
- Corning® UV-Transparent Microplates are ideal for determining protein and/or nucleic acid concentrations.
- Half Area microplate with UV-transparent well bottom.
- Free of DNase- and RNase.
- Acrylic base for greater strength and maximum leak resistance.
- Well volume of 205 μL; working volumes of 25 to 100 μL.
- 1cm pathlength corresponds to 175 μL volume.
- Lids are available separately.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service