Skip to Content
Merck
All Photos(1)

Key Documents

926752

Sigma-Aldrich

Sulfur dioxide solution

1.0 M in dichloromethane

Synonym(s):

Sulfur superoxide (6CI), Sulfurous acid anhydride, Sulfurous oxide

Sign Into View Organizational & Contract Pricing

Select a Size

25 ML
€240.00
100 ML
€575.00

€240.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 ML
€240.00
100 ML
€575.00

About This Item

Linear Formula:
SO2
CAS Number:
MDL number:
UNSPSC Code:
12352303
NACRES:
NA.06

€240.00


Please contact Customer Service for Availability

Request a Bulk Order

form

liquid

Quality Level

concentration

1.0 M in dichloromethane

density

1.307 g/mL

SMILES string

[S](=O)=O

InChI

1S/O2S/c1-3-2

InChI key

RAHZWNYVWXNFOC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Valuable reagent offered as a solution in dichloromethane for more convenient handling.

related product

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Carc. 2 - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service