Skip to Content
Merck
All Photos(1)

Key Documents

921009

Sigma-Aldrich

Silver(I) carbonate

≥99.99% trace metals basis

Synonym(s):

Silver carbonate

Sign Into View Organizational & Contract Pricing

Select a Size

50 G
€401.00

€401.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
50 G
€401.00

About This Item

Linear Formula:
Ag2CO3
CAS Number:
Molecular Weight:
275.75
Beilstein:
6936654
MDL number:
UNSPSC Code:
12352302
NACRES:
NA.23

€401.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥99.99% trace metals basis

mp

210 °C (dec.) (lit.)

density

6.08 g/mL at 25 °C (lit.)

SMILES string

[Ag]OC(=O)O[Ag]

InChI

1S/CH2O3.2Ag/c2-1(3)4;;/h(H2,2,3,4);;/q;2*+1/p-2

InChI key

KQTXIZHBFFWWFW-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Silver carbonate can be used in preparing silver-based photocatalysts with enhanced photodegradation properties.[1][2][3][4] Silver nanoparticles synthesized from Ag2CO3 in ionic liquid under microwave conditions is applicable in electrochemical CO2 reduction.[5]

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Silver carbonate loaded on activated carbon composite photocatalyst with enhanced photocatalytic activity under visible light irradiation.
Zhu X, et al.
Materials Technology: Advanced Performance Materials, 32(1), 38-45 (2017)
Hydrothermal synthesis of Ag2CO3-TiO2 loaded reduced graphene oxide nanocomposites with highly efficient photocatalytic activity.
Pant B, et al.
Chemical Engineering Communications , 207(5), 688-695 (2020)
The photocatalytic performance of silver halides − Silver carbonate heterostructures.
Dostanic J, et al.
Journal of Photochemistry and Photobiology A: Chemistry, 336, 1-7 (2017)
Synthesis of Silver Nanocatalysts Using Microwave Irradiation in Ionic Liquid for Reduction of Carbon Dioxide to CO By Solar-Driven Electrochemical.
Park J C, et al.
Electron Microscopy Reviews, 63, 3224-3224 null

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service