Key Documents
918016
Sigma-Aldrich
Air adaptor
for 3 mL Cartridge
Select a Size
1 EACH
€78.50
€78.50
Please contact Customer Service for Availability
Request a Bulk Order
Select a Size
Change View
1 EACH
€78.50
About This Item
UNSPSC Code:
41106401
NACRES:
NA.23
€78.50
Please contact Customer Service for Availability
Request a Bulk Order
description
Air adaptor, for 3 mL Cartridge, 1 EA
Quality Level
Application
The air adaptor connects cartridge to the air output of a fluid dispenser unit, such as a 3D bioprinter. Each adapter assembly is precision molded for secure attachment to the cartridge with minimal effort, making its connection to the air output of a fluid dispensing unit fast and seamless
Choose from one of the most recent versions:
Certificates of Analysis (COA)