Skip to Content
Merck
All Photos(1)

Key Documents

913448

Sigma-Aldrich

NanoFabTx reagent

Stabilizer-P

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
€63.60

€63.60


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 G
€63.60

About This Item

Empirical Formula (Hill Notation):
C6H10O2·C2H4O)x·CH4O
UNSPSC Code:
12162002
NACRES:
NA.23

€63.60


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

application(s)

advanced drug delivery

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

NanoFabTx reagent, Stabilizer-P is a component of our NanoFabTx drug formulation kits (920436, 915408), which include additional reagents and protocols for nanoprecipitation and microfluidics to synthesize 100-200 nm nanoparticles. NanoFabTx reagent, Stabilizer-P is a polymer stabilizer.

Application

NanoFabTx reagent, Stabilizer-P can be used as a refill for the component in our NanoFabTx drug formulation kits (920436, 915408).

Legal Information

NANOFABTX is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service