Skip to Content
Merck
All Photos(1)

Key Documents

86705

Sigma-Aldrich

Tetraethylurea

≥98.0%

Synonym(s):

TEU

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)2NCON(C2H5)2
CAS Number:
Molecular Weight:
172.27
Beilstein:
1763126
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

≥98.0%

refractive index

n20/D 1.447

bp

211-213 °C (lit.)

density

0.907 g/mL at 20 °C (lit.)

SMILES string

CCN(CC)C(=O)N(CC)CC

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

177.8 °F - closed cup

Flash Point(C)

81 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service