Skip to Content
Merck
All Photos(1)

Key Documents

481777

Sigma-Aldrich

Hexafluorotitanic acid solution

60 wt. % in H2O, 99.9% trace metals basis

Synonym(s):

Dihydrogen hexafluorotitanate(2−) , Fluorotitanic acid, Hexafluorotitanic acid, Hexafluorotitanic acid (H2 TiF6 )

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
€49.30

€49.30


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
500 G
€49.30

About This Item

Linear Formula:
H2TiF6
CAS Number:
Molecular Weight:
163.87
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.23

€49.30


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

99.9% trace metals basis

concentration

60 wt. % in H2O

density

1.675 g/mL at 25 °C

SMILES string

F.F.F[Ti](F)(F)F

InChI

1S/6FH.Ti/h6*1H;/q;;;;;;+4/p-4

InChI key

PFSXARRIPPWGNC-UHFFFAOYSA-J

Packaging

Packaged in poly bottles

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

"Deposition of high dielectric barium-doped titanium silicon oxide films on silicon using hexafluorotitanic acid and barium nitrate.
Lee MK, et al.
Electrochemical and Solid-State Letters, 7(11), B42-B44 (2004)
Characterization of Titanium Oxide Films Prepared by Liquid Phase Deposition Using Hexafluorotitanic Acid
Lee MK and Lei BH
Jpn J. App. Phys., Part I, 39(2), 103-103 null

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service