Skip to Content
Merck
All Photos(1)

Documents

861871

Sigma-Aldrich

Dexamethasone

98%

Synonym(s):

(11β,16α)-9-Fluoro-11,17,21-trihydroxy-16-methylpregna-1,4-diene-3,20-dione, 9α-Fluoro-16α-methyl-11β,17α,21-trihydroxy-1,4-pregnadiene-3,20-dione, 9α-Fluoro-16α-methylprednisolone, Prednisolone F

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H29FO5
CAS Number:
Molecular Weight:
392.46
Beilstein:
2066651
EC Number:
MDL number:
UNSPSC Code:
12352200

Assay

98%

optical activity

[α]25/D +77.5° in dioxane

mp

262-264 °C (lit.)

SMILES string

C[C@@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]2(C)[C@@]1(O)C(=O)CO

InChI

1S/C22H29FO5/c1-12-8-16-15-5-4-13-9-14(25)6-7-19(13,2)21(15,23)17(26)10-20(16,3)22(12,28)18(27)11-24/h6-7,9,12,15-17,24,26,28H,4-5,8,10-11H2,1-3H3/t12-,15+,16+,17+,19+,20+,21+,22+/m1/s1

InChI key

UREBDLICKHMUKA-CXSFZGCWSA-N

Gene Information

human ... NR3C1(2908)

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Glucocorticoid anti-inflammatory agent. Regulates T cell survival, growth, and differentiation. Inhibits the induction of nitric oxide synthase.

replaced by

Product No.
Description
Pricing

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aurélie Hautefort et al.
Chest, 147(6), 1610-1620 (2014-11-28)
Inflammation may contribute to the pathobiology of pulmonary arterial hypertension (PAH). Deciphering the PAH fingerprint on the inflammation orchestrated by dendritic cells (DCs) and T cells, key driver and effector cells, respectively, of the immune system, may allow the identification
Erich Piovan et al.
Cancer cell, 24(6), 766-776 (2013-12-03)
Glucocorticoid resistance is a major driver of therapeutic failure in T cell acute lymphoblastic leukemia (T-ALL). Here, we identify the AKT1 kinase as a major negative regulator of the NR3C1 glucocorticoid receptor protein activity driving glucocorticoid resistance in T-ALL. Mechanistically
S Choi et al.
British journal of anaesthesia, 112(3), 427-439 (2014-01-15)
Brachial plexus nerve blocks (BPBs) have analgesic and opioid sparing benefits for upper extremity surgery. Single-injection techniques are limited by the pharmacological duration and therapeutic index of local anaesthetics (LAs). Continuous catheter techniques, while effective can present management challenges. Off-label
Michael J White et al.
Cell, 159(7), 1549-1562 (2014-12-20)
Activated caspases are a hallmark of apoptosis induced by the intrinsic pathway, but they are dispensable for cell death and the apoptotic clearance of cells in vivo. This has led to the suggestion that caspases are activated not just to kill
A Keith Stewart et al.
The New England journal of medicine, 372(2), 142-152 (2014-12-09)
Lenalidomide plus dexamethasone is a reference treatment for relapsed multiple myeloma. The combination of the proteasome inhibitor carfilzomib with lenalidomide and dexamethasone has shown efficacy in a phase 1 and 2 study in relapsed multiple myeloma. We randomly assigned 792

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service