Skip to Content
Merck
All Photos(1)

Key Documents

83062

Sigma-Aldrich

tert-Butyl 12-hydroxy-4,7,10-trioxadodecanoate

≥97.0% (GC)

Synonym(s):

tert-Butyl 3-[2-(2-(2-hydroxyethoxy)ethoxy)ethoxy]propionate, tert-Butyl triethylene glycol-O-propionate

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€253.00

€253.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€253.00

About This Item

Empirical Formula (Hill Notation):
C13H26O6
CAS Number:
Molecular Weight:
278.34
Beilstein:
7638988
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€253.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

≥97.0% (GC)

form

liquid

refractive index

n20/D 1.446

density

1.052 g/mL at 20 °C (lit.)

functional group

ester
ether
hydroxyl

SMILES string

CC(C)(C)OC(=O)CCOCCOCCOCCO

InChI

1S/C13H26O6/c1-13(2,3)19-12(15)4-6-16-8-10-18-11-9-17-7-5-14/h14H,4-11H2,1-3H3

InChI key

KSXVEOLRERRELV-UHFFFAOYSA-N

Other Notes

Intermediate in the synthesis of a spacer for combinatorial chemistry[1]

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Angewandte Chemie (International Edition in English), 113, 379-379 (2001)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service