Skip to Content
Merck
All Photos(1)

Key Documents

40421

Sigma-Aldrich

1,11-Diamino-3,6,9-trioxaundecane

≥98.0% (GC), for peptide synthesis

Synonym(s):

2,2′-[Oxybis(ethylenoxy)]diethylamine

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€346.00
500 MG
€828.00

€346.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
€346.00
500 MG
€828.00

About This Item

Empirical Formula (Hill Notation):
C8H20N2O3
CAS Number:
Molecular Weight:
192.26
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

€346.00


Please contact Customer Service for Availability

Request a Bulk Order

Product Name

1,11-Diamino-3,6,9-trioxaundecane, ≥98.0% (GC)

Assay

≥98.0% (GC)

reaction suitability

reagent type: cross-linking reagent

SMILES string

NCCOCCOCCOCCN

InChI

1S/C8H20N2O3/c9-1-3-11-5-7-13-8-6-12-4-2-10/h1-10H2

InChI key

NIQFAJBKEHPUAM-UHFFFAOYSA-N

Application

Hydrophilic, homo-bifunctional linker, i.e. for the prep. of biotinylation reagents[1], for the prep. of dendrimers[2] and for the prep. of macrocycles[3].

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W Zhang et al.
Journal of the American Chemical Society, 123(37), 8914-8922 (2001-09-13)
An orthogonal, convergent route for the introduction of substoichiometric numbers of latent surface sites into dendrimers based on melamine is used to prepare targets that display one or two Boc-protected amines on the periphery. Asymmetry is the result of the
Ming Zeng et al.
ACS applied materials & interfaces, 11(34), 30661-30672 (2019-08-08)
Recessive dystrophic epidermolysis bullosa (RDEB) is a severe congenital skin fragility disease caused by COL7A1 mutations that result in type VII collagen (C7) deficiency. Herein, we report a synergistic polyplex system that can efficiently restore C7 expression in RDEB keratinocytes.
Dietrich, B., et al.
Tetrahedron, 29 , 1629-1645 (1973)
Kisuk Yang et al.
Biomaterials, 63, 177-188 (2015-06-27)
Paracrine signals produced from stem cells influence tissue regeneration by inducing the differentiation of endogenous stem or progenitor cells. However, many recent studies that have investigated paracrine signaling of stem cells have relied on either two-dimensional transwell systems or conditioned
Katherine D McReynolds et al.
Bioorganic & medicinal chemistry, 10(3), 625-637 (2002-01-30)
Interactions of recombinant gp120 (rgp120) with non-natural glycosphingolipids (GSLs) and structurally simpler analogues have been studied using a competitive adhesion assay. Conjugates of cellobiosyl ceramide and melibiosyl ceramide were synthetically prepared as water-soluble GSL analogues. These ligands were screened against

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service