Skip to Content
Merck
All Photos(2)

Key Documents

803871

Sigma-Aldrich

Mattson Boronate Urea Pinacol Ester

greener alternative
Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H21BF6N2O3
CAS Number:
Molecular Weight:
474.20
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

reaction suitability

reagent type: catalyst

greener alternative product characteristics

Catalysis
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

mp

230.39 °C

greener alternative category

storage temp.

2-8°C

SMILES string

CC(O1)(C)C(C)(C)OB1C2=CC=CC=C2NC(NC3=CC(C(F)(F)F)=CC(C(F)(F)F)=C3)=O

InChI

1S/C21H21BF6N2O3/c1-18(2)19(3,4)33-22(32-18)15-7-5-6-8-16(15)30-17(31)29-14-10-12(20(23,24)25)9-13(11-14)21(26,27)28/h5-11H,1-4H3,(H2,29,30,31)

InChI key

OJFYKTGJRXTBRH-UHFFFAOYSA-N

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for catalytic efficiency. Click here for more information.

Application

1-(3,5-bis(trifluoromethyl)phenyl)-3-(2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)urea is a user-friendly, bench stable white solid. When compared to conventional ureas, this urea is moderately acidic (pKa(DMSO) = 9.5) and benefits from improved catalytic abilities and stability in select reactions.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tyler J Auvil et al.
Angewandte Chemie (International ed. in English), 52(43), 11317-11320 (2013-09-17)
It takes two: A unique organocatalyzed cascade for the unsymmetric double arylation of α-nitrodiazoesters is described. This organocascade features the strategic use of carbene-activating anilines in conjunction with a urea catalyst, thus allowing for the synthesis of pharmaceutically attractive α-diarylesters
David M Nickerson et al.
Chemical communications (Cambridge, England), 49(39), 4289-4291 (2012-12-01)
The strategic incorporation of internal Lewis acids onto urea scaffolds gives rise to a family of tunable hydrogen bond donor catalysts. The nature of the Lewis acid and associated ligands affects the urea polarization, acidity, and activity in reactions of
Sonia S So et al.
Organic letters, 14(2), 444-447 (2012-01-03)
Boronate ureas operate as catalysts for the activation of nitrocyclopropane carboxylates in nucleophilic ring-opening reactions. A variety of amines were found to open the urea-activated nitrocyclopropane carboxylates, generating highly useful nitro ester building blocks in good yields. Standard manipulations allow
Andrea M Hardman et al.
Organic & biomolecular chemistry, 11(35), 5793-5797 (2013-08-03)
Highly functionalized oxazinanes are efficiently prepared through urea-catalyzed formal [3 + 3] cycloaddition reactions of nitrones and nitrocyclopropane carboxylates. The reaction system is general with respect to both the nitrocyclopropane carboxylates and nitrones enabling the preparation of a large family
Sonia S So et al.
Organic letters, 13(4), 716-719 (2011-01-14)
Boronate ureas are introduced as a new class of noncovalent catalysts for conjugate addition reactions with enhanced activity. Through intramolecular coordination of the urea functionality to a strategically placed Lewis acid, rate enhancements up to 10 times that of more

Related Content

Boronate ureas benefit from internal Lewis acid coordination of the urea cabonyl oxygen and the strategically placed boron. As a result of this structural feature, boronate ureas can be rendered more acidic than conventional urea hydrogen bond donor catalysts.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service