Anita Mattson – Professor Product Portal
Professor Anita Mattson
Boronate ureas benefit from internal Lewis acid coordination of the urea cabonyl oxygen and the strategically placed boron. As a result of this structural feature, boronate ureas can be rendered more acidic than conventional urea hydrogen bond donor catalysts. These more acidic boronate ureas are stable, storable, easy to use, and can enable enhanced and/or unique reactivity patterns when compared to conventional urea catalysts.
Mattson Group Website
Recent papers from the Mattson Group
Network error: Failed to fetch
DISCIPLINES
- Organic Synthetic Chemistry
- Catalysis