Skip to Content
Merck
All Photos(1)

Key Documents

794392

Sigma-Aldrich

2-Bromo-3-hexyl-5-iodothiophene solution

1.0 M in THF

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H14BrIS
Molecular Weight:
373.09
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

form

liquid

concentration

1.0 M in THF

refractive index

n20/D 1.449

density

1.085 g/mL at 25 °C

SMILES string

BrC1=C(CCCCCC)C=C(I)S1

InChI

1S/C10H14BrIS/c1-2-3-4-5-6-8-7-9(12)13-10(8)11/h7H,2-6H2,1H3

InChI key

SDMKEQYHISDGKT-UHFFFAOYSA-N

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 4 - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

1.4 °F

Flash Point(C)

-17 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service