Skip to Content
Merck
All Photos(2)

Key Documents

189871

Sigma-Aldrich

Ethylmagnesium bromide solution

3.0 M in diethyl ether

Synonym(s):

EtMgBr solution

Sign Into View Organizational & Contract Pricing

Select a Size

4 X 25 ML
€81.20
100 ML
€85.50
800 ML
€335.00
8 L
€1,950.00

€81.20


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
4 X 25 ML
€81.20
100 ML
€85.50
800 ML
€335.00
8 L
€1,950.00

About This Item

Linear Formula:
CH3CH2MgBr
CAS Number:
Molecular Weight:
133.27
Beilstein:
3587203
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

€81.20


Please contact Customer Service for Availability

Request a Bulk Order

form

liquid

Quality Level

reaction suitability

reaction type: Grignard Reaction

concentration

3.0 M in diethyl ether

bp

34.6 °C/1.013  hPa

density

1.02 g/mL at 25 °C

SMILES string

[CC[Mg]Br]
CC[Mg]Br

InChI

[1S/C2H5.BrH.Mg/c1-2;;/h1H2,2H3;1H;/q;;+1/p-1]
1S/C2H5.BrH.Mg/c1-2;;/h1H2,2H3;1H;/q;;+1/p-1

InChI key

[HSNUIYJWTSJUMS-UHFFFAOYSA-N]
TWTWFMUQSOFTRN-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Ethylmagnesium bromide, an organomagnesium compound, is commonly known as Grignard reagent.[1] It is a powerful carbon nucleophile mainly used for C-C bond formation.[2]

Application

Ethylmagnesium bromide solution (3.0 M in diethyl ether) can be used for copper(I)-catalyzed allylic substitution reaction.[2]

Packaging

The 25 mL Sure/Seal bottle is recommended as a single-use bottle. Repeated punctures will likely result in decreased performance of product.

Legal Information

Sure/Seal is a trademark of Sigma-Aldrich Co. LLC

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 1 - Skin Corr. 1B - STOT SE 3 - Water-react 1

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 1

Flash Point(F)

-40.0 °F - closed cup

Flash Point(C)

-40 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enantioselective Copper-Catalyzed SN2? Substitution with Grignard Reagents.
Alexakis A, et al.
Synlett, 0927-0930 (2001)
Organic Chemistry, 542-542 (2008)
Janne E Tønder et al.
Organic & biomolecular chemistry, 2(10), 1447-1455 (2004-05-12)
Copper mediated allylic substitutions and conjugate additions to geranyl, cinnamyl and allylic indole compounds have been investigated with the aim of finding a method for the creation of the all-carbon quaternary centre present in the natural product lyngbyatoxin A. Reaction
M Wältermann et al.
Microbiology (Reading, England), 146 ( Pt 5), 1143-1149 (2000-06-01)
The triacylglycerol (TAG)-accumulating, hydrocarbon-degrading bacterium Rhodococcus opacus strain PD630 and chemically induced storage-deficient mutants derived from this strain were investigated for their capability to accumulate storage lipids in the cytoplasm during cultivation under nitrogen-limiting conditions. Acylglycerols were analysed by matrix-associated
T Nishiyama et al.
Chemical & pharmaceutical bulletin, 48(12), 1999-2002 (2001-01-06)
The direct alpha-bromination of various ketones using trifluoromethanesulfonic anhydride and Grignard reagent or magnesium bromide in ether gave the corresponding alpha-bromo ketones in moderate to good yields under mild reaction conditions.

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service