Skip to Content
Merck
All Photos(1)

Key Documents

756784

Sigma-Aldrich

(Acetonitrile)[2-dicyclohexylphosphino-2′,4′,6′-triisopropylbiphenyl]gold(I) hexafluoroantimonate

Synonym(s):

XPhos Au(MeCN)SbF6

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C35H52AuF6NPSb
CAS Number:
Molecular Weight:
950.48
UNSPSC Code:
12352300
PubChem Substance ID:

form

solid

reaction suitability

core: gold
reagent type: catalyst

mp

133-136 °C

SMILES string

CC#[N+][Au].F[Sb-](F)(F)(F)(F)F.CC(C)c1cc(C(C)C)c(c(c1)C(C)C)-c2ccccc2P(C3CCCCC3)C4CCCCC4

InChI

1S/C33H49P.C2H3N.Au.6FH.Sb/c1-23(2)26-21-30(24(3)4)33(31(22-26)25(5)6)29-19-13-14-20-32(29)34(27-15-9-7-10-16-27)28-17-11-8-12-18-28;1-2-3;;;;;;;;/h13-14,19-25,27-28H,7-12,15-18H2,1-6H3;1H3;;6*1H;/q;;+1;;;;;;;+5/p-6

InChI key

PVKYSBDDIOTPED-UHFFFAOYSA-H

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Carc. 2 - STOT RE 2 Oral

Target Organs

Liver,Kidney

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service