Skip to Content
Merck
All Photos(1)

Key Documents

734942

Sigma-Aldrich

2-Chloro-5-(trifluoromethyl)pyridine-3-carbonyl chloride

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H2Cl2F3NO
CAS Number:
Molecular Weight:
244.00
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.496

density

1.585 g/mL at 25 °C

functional group

acyl chloride
chloro
fluoro

storage temp.

−20°C

SMILES string

FC(F)(F)c1cnc(Cl)c(c1)C(Cl)=O

InChI

1S/C7H2Cl2F3NO/c8-5-4(6(9)14)1-3(2-13-5)7(10,11)12/h1-2H

InChI key

CEQQAPUZEUIPCH-UHFFFAOYSA-N

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Skin Corr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service