Skip to Content
Merck
All Photos(3)

Key Documents

683094

Sigma-Aldrich

HS157

HS157
1 of 1 reviewers received a sample product or took part in a promotion

Umicore, 97%

Synonym(s):

Bis(1,5-cyclooctadiene)diiridium(I) dichloride, [Ir(1,5-cod)Cl]2, 1,5-Cyclooctadiene-iridium(I) chloride dimer, Chloro(1,5-cyclooctadiene)iridium(I) dimer, Di-μ-chlorobis[(1,2,5,6-η)-1,5-cyclooctadiene]diiridium, Iridium(I) chloride 1,5-cyclooctadiene complex dimer, [Ir(1,5-cod)Cl]2, [Ir(cod)Cl]2

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
€212.00
2 G
€370.00
10 G
€1,770.00

€212.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
€212.00
2 G
€370.00
10 G
€1,770.00

About This Item

Empirical Formula (Hill Notation):
C16H24Cl2Ir2
CAS Number:
Molecular Weight:
671.70
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

€212.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

powder

reaction suitability

core: iridium
reaction type: C-H Activation
reagent type: catalyst

mp

205 °C (dec.) (lit.)

SMILES string

Cl[Ir].Cl[Ir].C1CC=CCCC=C1.C2CC=CCCC=C2

InChI

1S/2C8H12.2ClH.2Ir/c2*1-2-4-6-8-7-5-3-1;;;;/h2*1-2,7-8H,3-6H2;2*1H;;/q;;;;2*+1/p-2/b2*2-1-,8-7-;;;;

InChI key

ZFOUDQNHNLDNLD-MIXQCLKLSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Umicore Precatalysts for Asymmetric and Cross-Coupling Catalysis

Metal precursor for asymmetric allylic substitutions.[1]

Legal Information

Product of Umicore

Additional information available at www.pmc.umicore.com

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P A Marks et al.
Journal of the National Cancer Institute, 92(15), 1210-1216 (2000-08-03)
Histone deacetylase (HDAC) inhibitors have been shown to be potent inducers of growth arrest, differentiation, and/or apoptotic cell death of transformed cells in vitro and in vivo. One class of HDAC inhibitors, hydroxamic acid-based hybrid polar compounds (HPCs), induce differentiation

Articles

Arylboronic acids and esters, vital tools in chemical transformations, find extensive use, particularly in the Suzuki-Miyaura cross-coupling reaction.

Precatalysts for asymmetric catalysis offer batch-to-batch consistency in a range of catalytic reactions, supporting diverse research needs.

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

1 of 1 reviewers received a sample product or took part in a promotion

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service