Skip to Content
Merck
All Photos(1)

Documents

346721

Sigma-Aldrich

(1R,2R)-(−)-1,2-Diaminocyclohexane

98%

Synonym(s):

(1R)-trans-1,2-Cyclohexanediamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H10(NH2)2
CAS Number:
Molecular Weight:
114.19
Beilstein:
2801643
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

optical activity

[α]20/D −25°, c = 5 in 1 M HCl

SMILES string

N[C@@H]1CCCC[C@H]1N

InChI

1S/C6H14N2/c7-5-3-1-2-4-6(5)8/h5-6H,1-4,7-8H2/t5-,6-/m1/s1

InChI key

SSJXIUAHEKJCMH-PHDIDXHHSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

(1R,2R)-(-)-1,2-Diaminocyclohexane may be used to prepare:
  • Chiral C2-symmetric diphenylphosphoramide and diphenylthiophosphoramide ligands by reacting with diphenylphosphinic chloride and diphenylthiophosphinic chloride, respectively.
  • Bis-urea and the amino-thiourea ligands by reacting with isocyanates and isothiocyanates, respectively.
  • Biologically active chiral palladium(II) and platinum(II) complexes.
Versatile ligand for the formation of metal complexes. Used in the synthesis of chiral tropocoronands which have potential utility in asymmetric catalysis.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

158.0 °F - closed cup

Flash Point(C)

70 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C 2-Symmetric diphenylphosphoramide and diphenylthiophosphoramide derived from (1R, 2R)-1, 2-diaminocyclohexane as ligands for the titanium (IV) alkoxide-promoted addition of diethylzinc to aldehydes.
Shi M and Sui WS
Tetrahedron Asymmetry, 10(17), 3319-3325 (1999)
Chiral amino-urea derivatives of (1R, 2R)-1, 2-diaminocyclohexane as ligands in the ruthenium catalysed asymmetric reduction of aromatic ketones by hydride transfer.
Bied C, et al.
Tetrahedron Asymmetry, 12(2), 329-336 (2001)
Luka Ðorđević et al.
Nature communications, 9(1), 3442-3442 (2018-08-26)
The chirality of (nano)structures is paramount in many phenomena, including biological processes, self-assembly, enantioselective reactions, and light or electron spin polarization. In the quest for new chiral materials, metallo-organic hybrids have been attractive candidates for exploiting the aforementioned scientific fields.
Chiral palladium (II) and platinum (II) complexes of diaminocyclohexane: X-ray structures of (1R, 2R)-(-)-1, 2-diaminocyclohexane dihydrochloride and its corresponding oxalato platinum (II) complex.
Abu-Surrah AS, et al.
Polyhedron, 22(12), 1529-1534 (2003)
Mukaiyama, T.
Aldrichimica Acta, 29, 59-59 (1996)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service