Skip to Content
Merck
All Photos(2)

Key Documents

647195

Sigma-Aldrich

5-Cyanophthalide

97%

Synonym(s):

1,3-Dihydro-1-oxo-5-isobenzofurancarbonitrile, 5-Cyano-3H-isobenzofuranone

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€43.40
25 G
€159.00

€43.40


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€43.40
25 G
€159.00

About This Item

Empirical Formula (Hill Notation):
C9H5NO2
CAS Number:
Molecular Weight:
159.14
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€43.40


Please contact Customer Service for Availability

Request a Bulk Order

Assay

97%

form

solid

mp

201-205 °C (lit.)

functional group

ester
nitrile

SMILES string

O=C1OCc2cc(ccc12)C#N

InChI

1S/C9H5NO2/c10-4-6-1-2-8-7(3-6)5-12-9(8)11/h1-3H,5H2

InChI key

XEEGWTLAFIZLSF-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service