Skip to Content
Merck
All Photos(2)

Key Documents

541087

Sigma-Aldrich

Ethyl 3,5-dihydroxybenzoate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(HO)2C6H3CO2CH2CH3
CAS Number:
Molecular Weight:
182.17
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

127-130 °C (lit.)

functional group

ester

SMILES string

CCOC(=O)c1cc(O)cc(O)c1

InChI

1S/C9H10O4/c1-2-13-9(12)6-3-7(10)5-8(11)4-6/h3-5,10-11H,2H2,1H3

InChI key

APHYVLPIZUVDTK-UHFFFAOYSA-N

General description

Ethyl 3,5-dihydroxybenzoate is a dihydroxybenzoic ester. It can be synthesized from 3,5-dihydroxybenzoic acid by esterification process with absolute ethanol.

Application

Ethyl 3,5-dihydroxybenzoate may be used as a starting reagent in the synthesis of ethyl 3,5-bis(ω-hydroxyoligo(ethyleneoxy))benzoates and p-alkoxycarbonylated palladium bis(phosphinite) PCP pincer complexes.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Structural variants of hyperbranched polyesters.
Kumar A and Ramakrishnan S.
Macromolecules, 29(7), 2524-2530 (1996)
PCP-Bis (phosphinite) pincer complexes: new homogeneous catalysts for a-arylation of ketones.
Churruca F, et al.
Tetrahedron Letters, 47(19), 3233-3237 (2006)
José Antonio Curiel et al.
Journal of agricultural and food chemistry, 57(14), 6224-6230 (2009-07-16)
Tannase is an enzyme with important biotechnological applications in the food industry. Previous studies have identified the tannase encoding gene in Lactobacillus plantarum and also have reported the description of the purification of recombinant L. plantarum tannase through a protocol

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service