Key Documents
BASMW1085
Sigma-Aldrich
Non-Aqueous Reference Electrode
Synonym(s):
Reference Electrode for Non-Aqueous Solutions
Select a Size
1 EACH
€189.00
€189.00
Please contact Customer Service for Availability
Request a Bulk Order
Select a Size
Change View
1 EACH
€189.00
About This Item
UNSPSC Code:
12352300
NACRES:
NA.23
€189.00
Please contact Customer Service for Availability
Request a Bulk Order
material
PTFE cap
glass body
description
frit tip with PTFE heat shrink
silver wire
Legal Information
Product of Bioanalytical Systems, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Questions
  1. Can I check the detailed specifications (thickness, usage environment, length, etc.) of this product?
    1 answer
    1. This is a product of Bioanalytical Systems, Inc. The length is 7.5 cm and the width at the tip of the probe is 5.7 mm. This electrode features a PTFE cap and glass body with a silver wire.
      Please see the supplier product image and drawing below.
      Helpful?