Skip to Content
Merck
All Photos(1)

Key Documents

540404

Sigma-Aldrich

1,1′-Bis(dimethylsilyl)ferrocene

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H22FeSi2
CAS Number:
Molecular Weight:
302.34
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
Product 540404 is not currently sold in your country. Contact Technical Service

Assay

97%

reaction suitability

core: iron
reagent type: catalyst
reaction type: Olefin Metathesis

refractive index

n20/D 1.564 (lit.)

bp

294-295 °C (lit.)

density

1.007 g/mL at 25 °C (lit.)

SMILES string

[Fe].[H][Si](C)(C)[C]1[CH][CH][CH][CH]1.[H][Si](C)(C)[C]2[CH][CH][CH][CH]2

InChI

1S/2C7H11Si.Fe/c2*1-8(2)7-5-3-4-6-7;/h2*3-6,8H,1-2H3;

InChI key

WAYRNCUBSTTWOE-UHFFFAOYSA-N

Application

  • Catalytic co-initiator in free radical photopolymerization and free radical promoted cationic polymerization reactions

Reactant for:
  • Hydrosilylataion and hydroboration reactions
  • Platinum/nickel catalyzed selective hydrosilylation of alkynes and alkenes
  • Iron-catalyzed dehydrogenative coupling reactions
  • Hydrosilylation and double silylation of carbonyl compounds

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

122.0 °F - closed cup

Flash Point(C)

50 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service