Skip to Content
Merck
All Photos(1)

Key Documents

525863

Sigma-Aldrich

trans-2-Aminocyclopentanol hydrochloride

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H11NO · HCl
CAS Number:
Molecular Weight:
137.61
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

191-196 °C (lit.)

functional group

hydroxyl

SMILES string

Cl.N[C@H]1CCC[C@@H]1O

InChI

1S/C5H11NO.ClH/c6-4-2-1-3-5(4)7;/h4-5,7H,1-3,6H2;1H/t4-,5-;/m0./s1

InChI key

ZFSXKSSWYSZPGQ-FHAQVOQBSA-N

General description

trans-2-Aminocyclopentanol hydrochloride is an aminocyclanol.[1] Its d,l-cis- and d,l-trans- forms have been synthesized.[2] The trans-form of the product can be produced in large (multigram) scale via carbamate addition protocol.[3] Cholinesterase inhibitory potential of cis-form of 2-aminocyclopentanol hydrochloride is higher (twofold) that of its trans-form.[1]

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

James A Birrell et al.
Organic letters, 15(12), 2895-2897 (2013-06-08)
A highly enantioselective addition of phenyl carbamate to meso-epoxides has been developed to efficiently generate protected trans-1,2-amino alcohols. This transformation is promoted by an oligomeric (salen)Co-OTf catalyst and has been used to prepare two useful 2-aminocycloalkanol hydrochlorides in enantiopure form
Stereochemistry of Aminocyclanols. Synthesis of cis Epimers via Oxazolines. The 2-Aminocyclopentanols*.
McCasland GE and Smith DA.
Journal of the American Chemical Society, 72(5), 2190-2195 (1950)
Preparation of antidotes for anticholinesterase poisoning. IV. Synthesis and protective effectiveness of 2'-(cis-and trans-2'-hydroxycyclohexyl) aminoethyl 1-phenylcyclopentanecarboxylate hydrochlorides.
Bannard RAB and Parkkari JH.
Canadian Journal of Chemistry, 48(9), 1377-1382 (1970)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service