Skip to Content
Merck
All Photos(1)

Documents

491616

Sigma-Aldrich

5-Fluorouracil-15N2

98 atom % 15N

Synonym(s):

5-fluoro-2,4(1H,3H)-Pyrimidinedione-1,3-15N2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H3F15N2O2
CAS Number:
Molecular Weight:
132.06
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

isotopic purity

98 atom % 15N

mp

282-286 °C (lit.)

mass shift

M+2

SMILES string

FC1=C[15NH]C(=O)[15NH]C1=O

InChI

1S/C4H3FN2O2/c5-2-1-6-4(9)7-3(2)8/h1H,(H2,6,7,8,9)/i6+1,7+1

InChI key

GHASVSINZRGABV-AKZCFXPHSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Karen Rossmassler et al.
Biodegradation, 30(2-3), 127-145 (2019-03-02)
Knowledge of the conditions that promote the growth and activity of pharmaceutical and personal care product (PPCP)-degrading microorganisms within mixed microbial systems are needed to shape microbiomes in biotreatment reactors and manage process performance. Available carbon sources influence microbial community

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service