Skip to Content
Merck
All Photos(1)

Key Documents

486310

Sigma-Aldrich

p-Xylene-(dimethyl-13C2)

99 atom % 13C

Synonym(s):

1,4-Dimethyl-13C2-benzene, p-Xylene-α,α-13C2

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€1,370.00

€1,370.00


Please contact Customer Service for Availability

Request more information

Select a Size

Change View
1 G
€1,370.00

About This Item

Linear Formula:
C6H4(13CH3)2
CAS Number:
Molecular Weight:
108.15
MDL number:
UNSPSC Code:
12191501
PubChem Substance ID:
NACRES:
NA.12

€1,370.00


Please contact Customer Service for Availability

Request more information

isotopic purity

99 atom % 13C

Quality Level

Assay

99% (CP)

refractive index

n20/D 1.495 (lit.)

bp

138 °C (lit.)

mp

12-13 °C (lit.)

density

0.882 g/mL at 25 °C

mass shift

M+2

SMILES string

[13CH3]c1ccc([13CH3])cc1

InChI

1S/C8H10/c1-7-3-5-8(2)6-4-7/h3-6H,1-2H3/i1+1,2+1

InChI key

URLKBWYHVLBVBO-ZDOIIHCHSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

77.0 °F - closed cup

Flash Point(C)

25.00 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service