Skip to Content
Merck
All Photos(1)

Documents

95755

Sigma-Aldrich

m-Xylylene diisocyanate

technical, ≥90% (GC)

Synonym(s):

1,3-Bis(isocyanatomethyl)benzene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H8N2O2
CAS Number:
Molecular Weight:
188.18
Beilstein:
781825
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical

Assay

≥90% (GC)

form

liquid

bp

88-90 °C/0.02 mmHg (lit.)

density

1.202 g/mL at 20 °C (lit.)

storage temp.

2-8°C

SMILES string

O=C=NCc1cccc(CN=C=O)c1

InChI

1S/C10H8N2O2/c13-7-11-5-9-2-1-3-10(4-9)6-12-8-14/h1-4H,5-6H2

InChI key

RTTZISZSHSCFRH-UHFFFAOYSA-N

Related Categories

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Aquatic Chronic 3 - Eye Dam. 1 - Flam. Liq. 3 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1A - STOT RE 1 Inhalation - STOT SE 1 Inhalation

Target Organs

Respiratory Tract

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

107.6 °F - closed cup

Flash Point(C)

42 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Suansuan Wang et al.
Soft matter, 13(45), 8609-8617 (2017-11-08)
A new class of bisurea derivatives bearing tetrahydroxy groups have been proven to be non-gelators in water and various organic solvents even under long-term sonication or efficient heating treatment. We found that it is possible to trigger physical gelation behaviour

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service