Skip to Content
Merck
All Photos(1)

Key Documents

45897

Sigma-Aldrich

3,7-Dimethyl-2,6-octadienyl acetate

mixture of isomers, >97.0% (sum of isomers, GC)

Sign Into View Organizational & Contract Pricing

Select a Size

500 ML
€42.20

€42.20


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
500 ML
€42.20

About This Item

Linear Formula:
CH3CO2CH2CH=C(CH3)CH2CH2CH=C(CH3)2
CAS Number:
Molecular Weight:
196.29
Beilstein:
1722815
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€42.20


Please contact Customer Service for Availability

Request a Bulk Order

Assay

>97.0% (sum of isomers, GC)

refractive index

n20/D 1.462

bp

236-242 °C (lit.)

density

0.913 g/mL at 20 °C (lit.)

functional group

ester

SMILES string

CC(=O)OC\C=C(/C)CC\C=C(/C)C

InChI

1S/C12H20O2/c1-10(2)6-5-7-11(3)8-9-14-12(4)13/h6,8H,5,7,9H2,1-4H3/b11-8+

InChI key

HIGQPQRQIQDZMP-DHZHZOJOSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bio-degradation of acetates of geraniol, nerol & citronellol by P. incognita: isolation & identification of metabolites.
K M Madyastha et al.
Indian journal of biochemistry & biophysics, 20(3), 136-140 (1983-06-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service