Skip to Content
Merck
All Photos(1)

Key Documents

33622

Sigma-Aldrich

(−)-O,O′-Dibenzoyl-L-tartaric acid monohydrate

≥99.0% (T)

Synonym(s):

(-)-Dibenzoyl-L-tartaric Acid Monohydrate

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
€35.30
500 G
€123.00

€35.30


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 G
€35.30
500 G
€123.00

About This Item

Empirical Formula (Hill Notation):
C18H14O8 · H2O
CAS Number:
Molecular Weight:
376.31
Beilstein:
4065263
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

€35.30


Please contact Customer Service for Availability

Request a Bulk Order

Assay

≥99.0% (T)

form

powder

optical activity

[α]20/D −110±3°, c = 5% in ethanol

mp

88-89 °C (lit.)

functional group

carboxylic acid
ester
phenyl

SMILES string

[H]O[H].OC(=O)[C@H](OC(=O)c1ccccc1)[C@@H](OC(=O)c2ccccc2)C(O)=O

InChI

1S/C18H14O8.H2O/c19-15(20)13(25-17(23)11-7-3-1-4-8-11)14(16(21)22)26-18(24)12-9-5-2-6-10-12;/h1-10,13-14H,(H,19,20)(H,21,22);1H2/t13-,14-;/m1./s1

InChI key

DXDIHODZARUBLA-DTPOWOMPSA-N

General description

(-)-O,O′-Dibenzoyl-L-tartaric acid monohydrate may be used as a chiral resolving agent for the resolution of racemic bioactive compounds such as (±)-3-(3-hydroxyphenyl)-N-(1-propyl)-piperidine[1] and (±)-2-(5,6-dimethoxy-1,2,3,4-tetrahydro-1-naphthyl)-imidazoline to isolate the corresponding (-)-enantiomeric forms.[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An efficient and inexpensive resolution of the potent dopaminergic substance 3-(3-Hydroxyphenyl)-N-(1-propyl)-piperidine (?)-3-PPP.
Law H, et al.
Tetrahedron Asymmetry, 2(10), 989-992 (1991)
Conformationally defined adrenergic agents. 5. Resolution, absolute configuration, and pharmacological characterization of the enantiomers of 2-(5, 6-dihydroxy-1, 2, 3, 4-tetrahydro-1-naphthyl) imidazoline: a potent agonist at. alpha.-adrenoceptors.
DeBernardis JF, et al.
Journal of Medicinal Chemistry, 30(6), 1011-1017 (1987)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service