Skip to Content
Merck
All Photos(1)

Key Documents

455865

Sigma-Aldrich

Hydrazine sulfate salt

99.999% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2NH2 · H2SO4
CAS Number:
Molecular Weight:
130.12
Beilstein:
8778859
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99.999% trace metals basis

form

solid

mp

254 °C (lit.)

SMILES string

NN.OS(O)(=O)=O

InChI

1S/H4N2.H2O4S/c1-2;1-5(2,3)4/h1-2H2;(H2,1,2,3,4)

InChI key

ZGCHATBSUIJLRL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Eye Dam. 1 - Skin Corr. 1 - Skin Sens. 1

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hepatic carcinogenesis in CBA-Cb-Se mice and Cb-Se rats by isonicotinic acid hydrazide and hydrazine sulfate.
L Severi et al.
Journal of the National Cancer Institute, 41(2), 331-349 (1968-08-01)
Fluorinated polyoxadiazole for high-temperature polymer electrolyte membrane fuel cells.
Dominique G and Nunes SP.
Journal of Membrane Science, 321(1), 114-122 (2008)
Tumorigenesis studies with 1,2-dimethylhydrazine dihydrochloride, hydrazine sulfate, and isonicotinic acid in golden hamsters.
B Toth
Cancer research, 32(4), 804-807 (1972-04-01)
Azo Compounds. 1 A Re-examination of the Structure of the Product from the Reaction of Heptane-2, 6-dione with Hydrazine Sulfate and Sodium Cyanide in Dilute Solution.
Overberger CG and Marks BS.
Journal of the American Chemical Society, 77(15), 4097-4100 (1955)
Ranendu Sekhar Das et al.
Dalton transactions (Cambridge, England : 2003), 42(11), 4068-4080 (2013-01-24)
PVP capped platinum nano particles (PNP) of 5 nm diameter were prepared and characterized as homogeneous and of spherical nature. At physiological pH range (6.0-8.0), these PNP catalyze the deoxygenation of phenoxazine group containing resazurin (1) by hydrazine. The observed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service