Skip to Content
Merck
All Photos(1)

Key Documents

1.04343

Supelco

Methenamine

GR for analysis Reag. Ph Eur

Synonym(s):

Methenamine, Hexamethylenetetramine, Hexamine, Formin, Urotropin

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
€59.60
500 G
€115.00
25 KG
€5,060.00

€59.60


Estimated to ship on01 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 G
€59.60
500 G
€115.00
25 KG
€5,060.00

About This Item

Empirical Formula (Hill Notation):
C6H12N4
CAS Number:
Molecular Weight:
140.19
MDL number:
UNSPSC Code:
12352005
EC Index Number:
202-905-8
NACRES:
NA.21
vapor pressure:
<0.01 hPa ( 20 °C)

€59.60


Estimated to ship on01 April 2025Details


Request a Bulk Order

Agency

reag. Ph. Eur.

Quality Level

vapor pressure

<0.01 hPa ( 20 °C)

Assay

≥99.5% (perchloric acid titration)

form

solid

autoignition temp.

390 °C

potency

9200 mg/kg LD50, oral (Rat)

dilution

(for analytical testing)

pH

7-10 (20 °C, 100 g/L in H2O)

transition temp

flash point 250 °C

density

1.33 g/cm3 at 20 °C

bulk density

600 kg/m3

storage temp.

15-25°C

SMILES string

N21CN3CN(C2)CN(C1)C3

InChI

1S/C6H12N4/c1-7-2-9-4-8(1)5-10(3-7)6-9/h1-6H2

InChI key

VKYKSIONXSXAKP-UHFFFAOYSA-N

Application


  • New phosphotetradecavanadate hybrids: crystal structure, DFT analysis, stability and binding interactions with bio-macromolecules.: This article discusses the synthesis and structural analysis of new hybrids involving methenamine, offering potential applications in catalysis and material science (Pessoa JC, 2022).

Analysis Note

Assay (perchloric acid titration): ≥ 99.5 %
Identity (IR-spectrum): passes test
Appearance: colorless or white to almost white crystalline powder
pH (100 g/l, water): 7.0 - 10.0
Chloride (Cl): ≤ 20 ppm
Sulphate (SO₄): ≤ 50 ppm
Heavy metals (as Pb): ≤ 10 ppm
NH₄ (Ammonium): ≤ 100 ppm
Sulfated ash (800 °C): ≤ 0.05 %

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Sol. 2 - Skin Sens. 1B

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 1

Flash Point(F)

482.0 °F - closed cup

Flash Point(C)

250 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service