Skip to Content
Merck
All Photos(1)

Documents

427373

Sigma-Aldrich

1,8-Diisocyanatooctane

98%

Synonym(s):

Octamethylene diisocyanate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
OCN(CH2)8NCO
CAS Number:
Molecular Weight:
196.25
Beilstein:
1772257
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

98%

form

liquid

refractive index

n20/D 1.455 (lit.)

bp

156 °C/15 mmHg (lit.)

density

1.007 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

O=C=NCCCCCCCCN=C=O

InChI

1S/C10H16N2O2/c13-9-11-7-5-3-1-2-4-6-8-12-10-14/h1-8H2

InChI key

QUPKOUOXSNGVLB-UHFFFAOYSA-N

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

228.2 °F - closed cup

Flash Point(C)

109 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vladimir Burmistrov et al.
Bioorganic & medicinal chemistry letters, 24(9), 2193-2197 (2014-04-02)
A series of inhibitors of the soluble epoxide hydrolase (sEH) containing two urea groups has been developed. Inhibition potency of the described compounds ranges from 2.0 μM to 0.4 nM. 1,6-(Hexamethylene)bis[(adamant-1-yl)urea] (3b) was found to be a potent slow tight

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service