Skip to Content
Merck
All Photos(1)

Key Documents

426237

Sigma-Aldrich

2,2,2,Trifluoroethanol-OD

99 atom % D

Synonym(s):

2,2,2-Trifluoroethan(ol-d), Trifluoroethyl alcohol-OD

Sign Into View Organizational & Contract Pricing

Select a Size

5 ML
€128.00
25 ML
€553.00

€128.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 ML
€128.00
25 ML
€553.00

About This Item

Linear Formula:
CF3CH2OD
CAS Number:
Molecular Weight:
101.05
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:
NACRES:
NA.21
technique(s):
NMR: suitable
bp:
77-80 °C (lit.)

€128.00


Please contact Customer Service for Availability

Request a Bulk Order

isotopic purity

99 atom % D

Quality Level

Assay

99% (CP)

form

liquid

technique(s)

NMR: suitable

refractive index

n20/D 1.3 (lit.)

bp

77-80 °C (lit.)

mp

−44 °C (lit.)

density

1.387 g/mL at 25 °C (lit.)

mass shift

M+1

SMILES string

[2H]OCC(F)(F)F

InChI

1S/C2H3F3O/c3-2(4,5)1-6/h6H,1H2/i6D

InChI key

RHQDFWAXVIIEBN-RAMDWTOOSA-N

General description

2,2,2-Trifluoroethan(ol-d) is a deuterated compound in which the hydrogen on the –OH group is substituted by deuterium.

Recommended products

Check out ChemisTwin®, our brand new online portal for identity confirmation and quantification of NMR spectra. Learn more or reach out to us for a free trial.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Dam. 1 - Flam. Liq. 3 - Repr. 1B - STOT RE 2 Inhalation

Target Organs

Blood

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

84.2 °F - closed cup

Flash Point(C)

29 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sara Suna Yücel et al.
Cell reports, 26(11), 3087-3099 (2019-03-14)
Unspliced XBP1 mRNA encodes XBP1u, the transcriptionally inert variant of the unfolded protein response (UPR) transcription factor XBP1s. XBP1u targets its mRNA-ribosome-nascent-chain-complex to the endoplasmic reticulum (ER) to facilitate UPR activation and prevents overactivation. Yet, its membrane association is controversial.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service