Skip to Content
Merck
All Photos(2)

Key Documents

424668

Sigma-Aldrich

1-(o-Tolyl)biguanide

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4NHC(=NH)NHC(=NH)NH2
CAS Number:
Molecular Weight:
191.23
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

powder

mp

143-145 °C (lit.)

SMILES string

Cc1ccccc1NC(=N)NC(N)=N

InChI

1S/C9H13N5/c1-6-4-2-3-5-7(6)13-9(12)14-8(10)11/h2-5H,1H3,(H6,10,11,12,13,14)

InChI key

SQZCAOHYQSOZCE-UHFFFAOYSA-N

Application

1-(o-Tolyl)biguanide was used as a ligand in the synthesis of new vanadyl complexes with the formula [VOL2].xH2O (HL=1-(o-tolyl)biguanide, x=0.5).

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bacalum E, et al.
Rev. Roum. Chim., 57(4-5), 427-434 (2012)
Aqueous Solution Studies of o-Tolyl Biguanide Complexes of Cobalt (II), Copper (II), and Nickel (II).
Siefker JR and Jardine LJ.
Proceedings of the Indiana Academy of Sciences, 77 (1967)
o-Tolyl Biguanide Complexes of Some Transition Metal Ions in 1-Methyl-2-Pyrrolidinone.
Siefker JR and Wence RL.
Proceedings of the Indiana Academy of Sciences, 75 (1965)
New vanadyl complexes with metformin derivatives as potential insulin mimetic agents.
Olar R, et al.
Journal of Thermal Analysis and Calorimetry, 110(1), 257-262 (2012)
[Method of determining o-tolylbiguanide in water].
I S Dukhovnaia
Gigiena i sanitariia, (5)(5), 45-46 (1984-05-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service