Skip to Content
Merck
All Photos(1)

Key Documents

424102

Sigma-Aldrich

4-Nitrophenyl trifluoromethanesulfonate

99%

Synonym(s):

4-Nitrophenyl triflate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3SO3C6H4NO2
CAS Number:
Molecular Weight:
271.17
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

99%

form

solid

mp

52-55 °C (lit.)

functional group

fluoro
nitro
triflate

SMILES string

[O-][N+](=O)c1ccc(OS(=O)(=O)C(F)(F)F)cc1

InChI

1S/C7H4F3NO5S/c8-7(9,10)17(14,15)16-6-3-1-5(2-4-6)11(12)13/h1-4H

InChI key

NDTIXHNCNLKURN-UHFFFAOYSA-N

Application

4-Nitrophenyl trifluoromethanesulfonate (4-nitrophenyltriflate) may be used in the following studies:
  • As a triflating agent in the synthesis of aryl triflates.[1]
  • As a reagent in the synthesis of aryl nonaflate.[2]
  • As a reagent for palladium catalyzed coupling reactions with different substrates.[3][4]

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Regioselective palladium-catalyzed arylation of vinyl ethers with 4-nitrophenyl triflate. Control by addition of halide ions.
Andersson CM and Hallberg A.
The Journal of Organic Chemistry, 53(9), 2112-2114 (1988)
Palladium-catalyzed cross-coupling reaction of organoboron compounds with organic triflates.
Ohe T, et al.
The Journal of Organic Chemistry, 58(8), 2201-2208 (1993)
4-Nitrophenyltriflate as a new triflating agent.
Zhu J, et al.
Tetrahedron Letters, 38(7), 1181-1182 (1997)
A high throughput synthesis of aryl triflate and aryl nonaflate promoted by a polymer supported base (PTBD).
Boisnard S, et al.
Tetrahedron Letters, 40(42), 7469-7472 (1999)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service