Skip to Content
Merck
All Photos(1)

Documents

421685

Sigma-Aldrich

(S)-2-(Boc-amino)-3-phenyl-1-propanol

98%

Synonym(s):

(S)-(−)-2-(tert-Butoxycarbonylamino)-3-phenyl-1-propanol, N-(tert-Butoxycarbonyl)-L-phenylalaninol, Boc-L-phenylalaninol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2CH[NHCO2C(CH3)3]CH2OH
CAS Number:
Molecular Weight:
251.32
Beilstein:
3652004
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

optical activity

[α]20/D −27°, c = 1 in chloroform

optical purity

ee: 99% (GLC)

reaction suitability

reaction type: Boc solid-phase peptide synthesis

mp

94-96 °C (lit.)

application(s)

peptide synthesis

SMILES string

CC(C)(C)OC(=O)N[C@H](CO)Cc1ccccc1

InChI

1S/C14H21NO3/c1-14(2,3)18-13(17)15-12(10-16)9-11-7-5-4-6-8-11/h4-8,12,16H,9-10H2,1-3H3,(H,15,17)/t12-/m0/s1

InChI key

LDKDMDVMMCXTMO-LBPRGKRZSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Employed in the synthesis of biochemically active compounds. For references, see Aldrichimica Acta.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aldrichimica Acta, 28, 13-13 (1995)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service