Skip to Content
Merck
All Photos(1)

Key Documents

647055

Sigma-Aldrich

4,4-Diethoxy-N,N-dimethyl-1-butanamine

Synonym(s):

1-(N,N-Dimethylamino)-4,4-diethoxybutane, 4,4-Diethoxy-N,N-dimethylbutanamine, 4,4-Diethoxy-N,N-dimethylbutylamine, 4-Dimethylaminobutanal diethyl acetal, N,N-Dimethyl-4-aminobutanal diethyl acetal

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€94.70
25 G
€418.00

€94.70


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€94.70
25 G
€418.00

About This Item

Empirical Formula (Hill Notation):
C10H23NO2
CAS Number:
Molecular Weight:
189.30
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€94.70


Please contact Customer Service for Availability

Request a Bulk Order

grade

technical grade

Quality Level

refractive index

n20/D 1.421 (lit.)

density

0.844 g/mL at 25 °C (lit.)

functional group

amine
ether

storage temp.

2-8°C

SMILES string

CCOC(CCCN(C)C)OCC

InChI

1S/C10H23NO2/c1-5-12-10(13-6-2)8-7-9-11(3)4/h10H,5-9H2,1-4H3

InChI key

QKXMWBLNSPNBEY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

157.8 °F

Flash Point(C)

69.9 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service