Skip to Content
Merck
All Photos(2)

Key Documents

418625

Sigma-Aldrich

Tetrabutylammonium difluorotriphenylstannate

97%

Synonym(s):

Tetrabutylammonium difluorotriphenyltin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2CH2CH2)4N[(C6H5)3SnF2]
CAS Number:
Molecular Weight:
630.48
Beilstein:
8668854
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

192-193 °C (lit.)

functional group

amine

SMILES string

CCCC[N+](CCCC)(CCCC)CCCC.F[Sn-](F)(c1ccccc1)(c2ccccc2)c3ccccc3

InChI

1S/C16H36N.3C6H5.2FH.Sn/c1-5-9-13-17(14-10-6-2,15-11-7-3)16-12-8-4;3*1-2-4-6-5-3-1;;;/h5-16H2,1-4H3;3*1-5H;2*1H;/q+1;;;;;;+1/p-2

InChI key

ODMXVCNGZGLSRS-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Reactant for:
  • Reaction with K tert-butoxide to give disilylated fluorosilylenoid for subsequent self-condensation to fluorodisilanyl anion
  • Cross-coupling reactions with vinyl and aryl triflates
  • Cocatalyst in phase-transfer fluorination of alkyl halides and sulfonates
  • Used as a nucleophilic fluorinating agent

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service