Skip to Content
Merck
All Photos(2)

Documents

416673

Sigma-Aldrich

Bis(cyclopentadienyl)zirconium(IV) bis(trifluoromethanesulfonate)tetrahydrofuran complex

97%

Synonym(s):

Bis(cyclopentadienyl)zirconium(IV) bis(trifluoromethanesulfonate) THF complex, Di(cyclopentadienyl)zirconium(IV) bis(trifluoromethanesulfonate) THF complex, Zirconocene bis(trifluoromethanesulfonate) THF complex

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H10F6O6S2Zr · C4H8O
CAS Number:
Molecular Weight:
591.65
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

reaction suitability

core: zirconium
reagent type: catalyst

SMILES string

[Zr++].[CH]1[CH][CH][CH][CH]1.[CH]2[CH][CH][CH][CH]2.C3CCOC3.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F

InChI

1S/2C5H5.C4H8O.2CHF3O3S.Zr/c3*1-2-4-5-3-1;2*2-1(3,4)8(5,6)7;/h2*1-5H;1-4H2;2*(H,5,6,7);/q;;;;;+2/p-2

InChI key

OFTAVORSNHLQHT-UHFFFAOYSA-L

Application

Reagent for synthesis of β-hFSH with Chitobiose units

Reactant for polyoxometalate transfer for synthesis of zirconocene polyoxometalate

Transition metal based Lewis acid catalyst

Catalyst for:
  • Mukaiyama cross-aldol reactions
  • Diels-Alder reactions

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

174.2 °F - closed cup

Flash Point(C)

79 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service