Skip to Content
Merck
All Photos(2)

Key Documents

395420

Sigma-Aldrich

Zirconium(IV) chloride tetrahydrofuran complex (1:2)

99%

Synonym(s):

Tetrachlorobis(tetrahydrofuran)zirconium, Zirconium tetrachloride tetrahydrofuran complex

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€71.80

€71.80


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€71.80

About This Item

Linear Formula:
ZrCl4 · 2OC4H8
CAS Number:
Molecular Weight:
377.25
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
99%
form:
powder

€71.80


Please contact Customer Service for Availability

Request a Bulk Order

vapor pressure

152 mmHg ( 25 °C)(lit.)

Quality Level

Assay

99%

form

powder

reaction suitability

reagent type: catalyst
core: zirconium

mp

175-177 °C (lit.)

application(s)

battery manufacturing

storage temp.

2-8°C

SMILES string

Cl[Zr](Cl)(Cl)Cl.C1CCOC1.C2CCOC2

InChI

1S/2C4H8O.4ClH.Zr/c2*1-2-4-5-3-1;;;;;/h2*1-4H2;4*1H;/q;;;;;;+4/p-4

InChI key

VDJJKYYENYIHFF-UHFFFAOYSA-J

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Flam. Sol. 1 - Skin Corr. 1B

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service