Skip to Content
Merck
All Photos(1)

Key Documents

415367

Sigma-Aldrich

N-Ethyl-p-toluenesulfonamide

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4SO2NHC2H5
CAS Number:
Molecular Weight:
199.27
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

bp

208 °C/745 mmHg (lit.)

mp

63-65 °C (lit.)

SMILES string

CCNS(=O)(=O)c1ccc(C)cc1

InChI

1S/C9H13NO2S/c1-3-10-13(11,12)9-6-4-8(2)5-7-9/h4-7,10H,3H2,1-2H3

InChI key

OHPZPBNDOVQJMH-UHFFFAOYSA-N

General description

N-Ethyl-p-toluenesulfonamide is an herbicide. It is an organic contaminant present in the river Rhine. N-Ethyl-p-toluenesulfonamide/ N-bromo succinimmide (NBS) along with N-methyl-p-toluenesulfonamide/NBS serves as nitrogen/halogen resources during the synthesis of vicinal haloamino ketone derivatives. Reaction of epichlorohydrin with bisphenol is useful for the industrial preparation of epoxide resins.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

New Epoxide Resins by Reaction of Epichlorohydrin with Sulfonamides.
Cohen M.
Industrial & Engineering Chemistry Research, 47(10), 2095-2101 (1955)
G M Alink et al.
Mutation research, 631(2), 93-100 (2007-06-15)
Surface water used for drinking-water preparation requires continuous monitoring for the presence of toxic compounds. For monitoring of genotoxic compounds fish models have been developed, such as the Eastern mudminnow (Umbra pygmaea L.) because of its clearly visible 22 meta-centric
Hao Sun et al.
Chemical biology & drug design, 75(3), 269-276 (2010-03-25)
The combinations of N-methyl-p-toluenesulfonamide/NBS and N-ethyl-p-toluenesulfonamide/NBS were found to be good nitrogen/halogen resources for the aminohalogenation of alpha,beta-unsaturated ketones in the presence of Ni(OAc)(2) as the catalyst for the synthesis of vicinal haloamino ketone derivatives. The introduction of N-alkyl groups
L Kanerva et al.
Acta dermato-venereologica, 73(2), 126-129 (1993-04-01)
Dental personnel are exposed to many sensitizing compounds at work and often develop multiple delayed allergies. Here we report on a dentist who got sensitized to several products that have not, or only seldom, caused sensitization earlier. These products were:
Ingun Skjevrak et al.
Food additives and contaminants, 22(10), 1012-1022 (2005-10-18)
A procedure used by the Norwegian Food Safety Authority for surveillance of contaminants from plastic food contact materials (polyolefin drinking bottles, water boilers, polyamide cooking utensils and plastic multi-layer materials) is described. It is based on gas chromatographic-mass spectrometric (GC/MS)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service