Skip to Content
Merck
All Photos(1)

Key Documents

365696

Sigma-Aldrich

4-Methoxyphenylacetyl chloride

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H4CH2COCl
CAS Number:
Molecular Weight:
184.62
Beilstein:
908061
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

refractive index

n20/D 1.54 (lit.)

bp

143 °C/10 mmHg (lit.)

density

1.208 g/mL at 25 °C (lit.)

SMILES string

COc1ccc(CC(Cl)=O)cc1

InChI

1S/C9H9ClO2/c1-12-8-4-2-7(3-5-8)6-9(10)11/h2-5H,6H2,1H3

InChI key

CXJOONIFSVSFAD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4-Methoxyphenylacetyl chloride may be used in the synthesis of:
  • substituted spiro[4.5]decane
  • 1-(4′-hydroxybenzyl)-6, 7-dihydroxy-1, 2, 3, 4-tetrahydroisoquinoline (higenamine), a cardiotonic principle of aconite root
  • 2,5-naphthyridine
  • (R)-(+)-nor-roefractine

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael A Letavic et al.
Bioorganic & medicinal chemistry letters, 17(9), 2566-2569 (2007-02-20)
A series of novel tetrahydronaphthyridine-based histamine H(3) ligands that have serotonin reuptake transporter inhibitor activity is described. The 1,2,3,4-tetrahydro-2,6-naphthyridine scaffold is assembled via the addition of a nitrostyrene to a metalated pyridine followed by reduction and cyclization to form the
Synthesis of substituted spiro [4.5] deca-3, 6, 9-triene-2, 8-diones: an expeditious route to the spiro [4.5] decane terpene skeleton.
Haack RA and Beck KR.
Tetrahedron Letters, 30(13), 16505-16508 (1989)
N Cabedo et al.
Journal of natural products, 61(6), 709-712 (1998-06-27)
(R)-(+)-nor-Roefractine (1) was synthesized by the Bischler-Napieralski route, using asymmetric reduction of the 1, 2-didehydro precursor imine with sodium (S)-N-CBZ-prolinyloxyborohydride. Compound 1 was able to displace [3H]-raclopride (a D2 dopamine receptor-selective ligand) from its specific binding sites in rat striatum
Synthesis of higenamine, A cardiotonic principle of aconite root.
Chang K-C, et al.
Archives of Pharmacal Research, 7(2), 133-136 (1984)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service