Skip to Content
Merck
All Photos(1)

Key Documents

348953

Sigma-Aldrich

Octyl cyanoacetate

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NCCH2CO2(CH2)7CH3
CAS Number:
Molecular Weight:
197.27
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

liquid

refractive index

n20/D 1.449 (lit.)

bp

95 °C/0.11 mmHg (lit.)

density

0.934 g/mL at 25 °C (lit.)

functional group

ester
nitrile

SMILES string

CCCCCCCCOC(=O)CC#N

InChI

1S/C11H19NO2/c1-2-3-4-5-6-7-10-14-11(13)8-9-12/h2-8,10H2,1H3

InChI key

MBCTWXWDQMXVDF-UHFFFAOYSA-N

Application

Octyl cyanoacetate may be used in the preparation of a small molecule with a dithienosilole core for the high efficiency solution-processed organic photovoltaic cells. It may be used in the preparation of small molecule DCAO3TBDT with the acceptor-donor-acceptor (A-D-A) structure and benzo[1,2-b:4,5-b′]dithiophene (BDT) as the central building block.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A planar small molecule with dithienosilole core for high efficiency solution-processed organic photovoltaic cells.
Zhou J, et al.
Chemistry of Materials, 23(21), 4666-4668 (2011)
Jiaoyan Zhou et al.
Journal of the American Chemical Society, 134(39), 16345-16351 (2012-09-20)
Small molecules, namely, DCAO(3)TBDT and DR(3)TBDT, with 2-ethylhexoxy substituted BDT as the central building block and octyl cyanoacetate and 3-ethylrhodanine as different terminal units with the same linkage of dioctyltertthiophene, have been designed and synthesized. The photovoltaic properties of these

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service