Skip to Content
Merck
All Photos(1)

Key Documents

341517

Sigma-Aldrich

Tetrahydro-2-furoic acid

97%

Synonym(s):

Tetrahydrofuran-2-carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H8O3
CAS Number:
Molecular Weight:
116.12
Beilstein:
109671
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

contains

250 ppm BHT as inhibitor

refractive index

n20/D 1.46 (lit.)

bp

128-129 °C/13 mmHg (lit.)
237-243 °C (lit.)

density

1.209 g/mL at 25 °C (lit.)

functional group

carboxylic acid
ether

SMILES string

OC(=O)C1CCCO1

InChI

1S/C5H8O3/c6-5(7)4-2-1-3-8-4/h4H,1-3H2,(H,6,7)

InChI key

UJJLJRQIPMGXEZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Tetrahydro-2-furoic acid inhibits the programmed cell death of Xanthomonas campestris strains.[1] It is a specific inhibitor of proline dehydrogenase, a mitochondrial flavoenzyme that oxidizes proline.[2]

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Surbhi Wadhawan et al.
PloS one, 9(5), e96423-e96423 (2014-05-03)
Xanthomonas campestris strains have been reported to undergo programmed cell death (PCD) in a protein rich medium. Protein hydrolysates used in media such as nutrient broth comprise of casein digest with abundance of proline and glutamate. In the current study
Navasona Krishnan et al.
Free radical biology & medicine, 44(4), 671-681 (2007-11-27)
The potential of proline to suppress reactive oxygen species (ROS) and apoptosis in mammalian cells was tested by manipulating intracellular proline levels exogenously and endogenously by overexpression of proline metabolic enzymes. Proline was observed to protect cells against H(2)O(2), tert-butyl
S Bando et al.
Acta crystallographica. Section C, Crystal structure communications, 46 ( Pt 1), 106-108 (1990-01-15)
C5H10O4, Mr = 134.31, monoclinic, P2(1), a = 11.670 (2), b = 6.177 (3), c = 10.704 (2) A, beta = 115.11 (2) degrees, V = 698.7 (4) A3, Z = 4, Dm = 1.26, Dx = 1.28 g cm-3
Chengwen Feng et al.
Animals : an open access journal from MDPI, 10(9) (2020-09-05)
Proline was reported to improve sperm quality in rams, stallions, cynomolgus monkeys, donkeys, and canines during cryopreservation. However, the underlying mechanism remains unclear. The aim of this study was to investigate the effect of proline on boar semen during liquid
Tatsuki Sato et al.
BioMed research international, 2020, 7245782-7245782 (2020-12-05)
Candida albicans undergoes a yeast-to-hyphal transition that has been recognized as a virulence property as well as a turning point leading to biofilm formation associated with candidiasis. It is known that yeast-to-hyphal transition is induced under complex environmental conditions including

Questions

  1. What is the solubility of item 341517, Tetrahydro-2-furoic acid, from Sigma-Aldrich, in water?

    1 answer
    1. The solubility of Tetrahydro-2-furoic acid in water is reported as 27.9 mg/ml or 0.24 mol/l.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service