Skip to Content
Merck
All Photos(1)

Key Documents

633658

Sigma-Aldrich

4-Thiazolecarboxylic acid

97%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€27.10
5 G
€112.00

€27.10


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€27.10
5 G
€112.00

About This Item

Empirical Formula (Hill Notation):
C4H3NO2S
CAS Number:
Molecular Weight:
129.14
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€27.10


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

solid

mp

195-199 °C (lit.)

functional group

carboxylic acid

SMILES string

OC(=O)c1cscn1

InChI

1S/C4H3NO2S/c6-4(7)3-1-8-2-5-3/h1-2H,(H,6,7)

InChI key

HMVYYTRDXNKRBQ-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chloe Dubreil et al.
Small (Weinheim an der Bergstrasse, Germany), 14(40), e1802053-e1802053 (2018-09-06)
Nanoparticle (NP) administration is among the most attractive approaches to exploit the synergy of different copackaged molecules for the same target. In this work, iron oxide NPs are surface-engineered for the copackaging of the autoantigen proinsulin, a major target of
Angela Rico de Souza et al.
Toxicological sciences : an official journal of the Society of Toxicology, 140(1), 204-223 (2014-04-23)
Cigarette smoke is associated with chronic and enhanced pulmonary inflammation characterized by increased cytokine production and leukocyte recruitment to the lung. Although the aryl hydrocarbon receptor (AhR) is well-known to mediate toxic effects of manmade environmental contaminants, the AhR has
Hideo Satsu et al.
Cytotechnology, 67(4), 621-632 (2014-03-29)
The aryl hydrocarbon receptor (AHR) is a ligand-dependent transcription factor. It heterodimerizes with aryl hydrocarbon nuclear translocator, binds to the xenobiotic-responsive element (XRE), and enhances the transcription of genes encoding xenobiotic metabolizing enzymes. AHR also plays important roles in the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service